accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOOOCH3

CAS RN 607-81-8

IUPAC Name: 1,3-diethyl 2-benzylpropanedioate
Synonyms: benzylmalonic acid, diethyl ester
Molecular Weight (g/mol): 250.29
Molecular Formula: C14H18O4
InChi Key: ICZLTZWATFXDLP-UHFFFAOYSA-N
SMILES: CCOC(=O)C(CC1=CC=CC=C1)C(=O)OCC
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results