accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 60835-69-0

IUPAC Name: 15-nitro-2,3,5,6,8,9,11,12-octahydro-1,4,7,10,13-benzopentaoxacyclopentadecine
Synonyms: 15-nitro-2,3,5,6,8,9,11,12-octahydro-1,4,7,10,13-benzopentaoxacyclopentadecine
Molecular Weight (g/mol): 313.31
Molecular Formula: C14H19NO7
InChi Key: XIWRBQVYCZCEPG-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2OCCOCCOCCOCCOC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results