accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 61-25-6

IUPAC Name: hydrogen 1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxyisoquinoline chloride
Synonyms: hydrogen papaverine chloride
Molecular Weight (g/mol): 375.85
Molecular Formula: C20H22ClNO4
InChi Key: UOTMYNBWXDUBNX-UHFFFAOYSA-N
SMILES: [H+].[Cl-].COC1=CC=C(CC2=C3C=C(OC)C(OC)=CC3=CC=N2)C=C1OC
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results