missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Papaverine Hydrochloride, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Papaverine Hydrochloride |
| CAS | 61-25-6 |
| Analyte or Component Names | Papaverine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H21 N O4 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Papaverine hydrochloride, 1-(3,4-Dimethoxybenzyl)-6,7-dimethoxyisoquinoline hydrochloride, Isoquinoline, 1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxy-, hydrochloride, Isoquinoline, 6,7-dimethoxy-1-veratryl-, hydrochloride, 1-(3,4-Dimethoxybenzyl)-6,7-dimethoxyisoquinoline hydrochloride, 6,7,3',4'-Tetramethoxy-1-benzylisoquinoline hydrochloride, 6,7-Dimethoxy-1-(3,4-dimethoxybenzyl)isoquinoline hydrochloride, 6,7-Dimethoxy-1-veratrylisoquinoline hydrochloride, Artegodan, Cardiospan, Cardoverina, Cepaverin, Cerebid, Cerespan, Dispamil, Drapavel, Dynovas, Myobid, Optenyl, Pameion, Panergon, Papavarine chlorhydrate, Papaverinium chloride, Papaversan, Paptial TR, Pavabid, Pavacap, Pavacen, Pavadel, Pavagen, Pavakey, Pavased, Paveron, Ro-Papav, Spasmo Nit, Therapav, Vasal, Vasorin, Vasospan, Noscapine Hydrochloride Hydrate Impurity A as Hydrochloride, Noscapine Impurity A as Hydrochloride, Noscapine Impurity A as Hydrochloride |
| InChI Formula | InChI=1S/C20H21NO4.ClH/c1-22-17-6-5-13(10-18(17)23-2)9-16-15-12-20(25-4)19(24-3)11-14(15)7-8-21-16;/h5-8,10-12H,9H2,1-4H3;1H |
| Solution Type | Neat |
| SMILES | Cl.COc1ccc(Cc2nccc3cc(OC)c(OC)cc23)cc1OC |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?