accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3C(S)NH2OHOCH3

CAS RN 61-90-5

IUPAC Name: 2-amino-4-methylpentanoic acid
Synonyms: leucine leuzin leucin Hleu
Molecular Weight (g/mol): 131.18
Molecular Formula: C6H13NO2
InChi Key: ROHFNLRQFUQHCH-UHFFFAOYNA-N
SMILES: CC(C)CC(N)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (22)
Quantity 
  • (2)
  • (1)
  • (8)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (3)
Grade 
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (7)
  • (3)
  • (3)
  • (6)
  • (2)
  • (2)
  • (3)
  • (22)
  • (2)
  • (1)
  • (8)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (4)
  • (2)