accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 610-92-4

IUPAC Name: 2,5-dihydroxybenzene-1,4-dicarboxylic acid
Synonyms: 2,5-dihydroxybenzene-1,4-dicarboxylic acid
Molecular Weight (g/mol): 198.13
Molecular Formula: C8H6O6
InChi Key: OYFRNYNHAZOYNF-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(O)=C(C=C1O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results