accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 6105-75-5

H3CH3CNH2NO

CAS RN 6105-75-5

IUPAC Name: 2-methyl-1-(morpholin-4-yl)propan-2-amine
Synonyms: 2-methyl-1-(morpholin-4-yl)propan-2-amine
Molecular Weight (g/mol): 158.25
Molecular Formula: C8H18N2O
InChi Key: JCXYZQUYVNLCTG-UHFFFAOYSA-N
SMILES: CC(C)(N)CN1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results