accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHOH(Z)

CAS RN 6117-80-2

IUPAC Name: (2Z)-but-2-ene-1,4-diol
Synonyms: 2-butene-1,4-diol (cis)
Molecular Weight (g/mol): 88.11
Molecular Formula: C4H8O2
InChi Key: ORTVZLZNOYNASJ-UPHRSURJSA-N
SMILES: OC\C=C/CO
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)