accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SOOOHHON(S)(R)(R)NH2CHOOH3CHHN(S)(R)(R)NH2CHOOH3CHHH2OH2O

CAS RN 6119-70-6

IUPAC Name: bis((R)-[(2S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol) sulfuric acid dihydrate
Synonyms: bis(quinine) sulfuric acid dihydrate
Molecular Weight (g/mol): 782.95
Molecular Formula: C40H54N4O10S
InChi Key: ZHNFLHYOFXQIOW-LRXYSQQVSA-N
SMILES: O.O.OS(O)(=O)=O.COC1=CC=C2N=CC=C([C@@H](O)[C@@H]3CC4CCN3C[C@@H]4C=C)C2=C1.COC1=CC=C2N=CC=C([C@@H](O)[C@@H]3CC4CCN3C[C@@H]4C=C)C2=C1
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)