accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CN

CAS RN 612-60-2

IUPAC Name: 7-methylquinoline
Synonyms: 7-methylquinoline
Molecular Weight (g/mol): 143.19
Molecular Formula: C10H9N
InChi Key: KDYVCOSVYOSHOL-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=CC=NC2=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)