missing translation for 'onlineSavingsMsg'
Learn More

m-Toluquinoline, TRC

Product Code. 30455310
Change view
Click to view available options
Quantity:
2.5 g
10 g
25 g
Unit Size:
10g
2.5g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30455310 25 g 25g
30451190 10 g 10g
30451853 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30455310 Supplier Toronto Research Chemicals Supplier No. TRCT53655525G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 612-60-2
Molecular Formula C10 H9 N
Molecular Weight (g/mol) 143.185
Synonym 7-Methylquinoline, NSC 65665, m-Toluquinoline
IUPAC Name 7-methylquinoline
SMILES Cc1ccc2cccnc2c1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C10H9N/c1-8-4-5-9-3-2-6-11-10(9)7-8/h2-7H,1H3
Formula Weight 143.0735
Chemical Name or Material 7-Methylquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.