accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 61295-50-9

H3COCH3SOH3C

CAS RN 61295-50-9

IUPAC Name: 4-[(2-methylfuran-3-yl)sulfanyl]nonan-5-one
Synonyms: 4-[(2-methylfuran-3-yl)sulfanyl]nonan-5-one
Molecular Weight (g/mol): 254.39
Molecular Formula: C14H22O2S
InChi Key: PEYZZTQOVLTVHN-UHFFFAOYNA-N
SMILES: CCCCC(=O)C(CCC)SC1=C(C)OC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results