missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Methyl-3-furylthio)-5-nonanone, 95%, Thermo Scientific™

Product Code. 11471860
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11471860 500 mg 500mg
11461860 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11471860 Supplier Thermo Scientific Alfa Aesar Supplier No. H27560.ME
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61295-50-9
Molecular Formula C14H22O2S
Molecular Weight (g/mol) 254.388
MDL Number MFCD08741453
InChI Key PEYZZTQOVLTVHN-UHFFFAOYSA-N
Synonym 4-2-methyl-3-furyl thio-5-nonanone, 4-2-methyl-3-furyl thio nonan-5-one, 4-2-methyl-3-furanyl thio-5-nonanone, 1,3-dipropylacetonyl 2-methyl-3-furyl sulfide, 4-2-methyl-furan-3-ylsulfanyl-nonan-5-one, 4-2-methyl-3-furylthio-5-nonanone, 4-2-methylfuran-3-yl sulfanylnonan-5-one, 5-nonanone, 4-2-methyl-3-furanyl thio, 4-2-methylfuran-3-yl sulfanyl nonan-5-one
PubChem CID 62182
IUPAC Name 4-(2-methylfuran-3-yl)sulfanylnonan-5-one
SMILES CCCCC(=O)C(CCC)SC1=C(OC=C1)C

Specifications

Refractive Index 1.4985
Quantity 500 mg
Formula Weight 254.39
Percent Purity 95%
Chemical Name or Material 4-(2-Methyl-3-furylthio)-5-nonanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.