accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 613-42-3

CAS RN 613-42-3

IUPAC Name: 4-benzyl-1,1'-biphenyl
Synonyms: 4-benzylbiphenyl
Molecular Weight (g/mol): 244.34
Molecular Formula: C19H16
InChi Key: AGPLQTQFIZBOLI-UHFFFAOYSA-N
SMILES: C(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results