accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 613-62-7

O

CAS RN 613-62-7

IUPAC Name: 2-(benzyloxy)naphthalene
Synonyms: 2-(benzyloxy)naphthalene
Molecular Weight (g/mol): 234.3
Molecular Formula: C17H14O
InChi Key: WLTCCDHHWYAMCG-UHFFFAOYSA-N
SMILES: C(OC1=CC=C2C=CC=CC2=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results