accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

KH3COOOHO

CAS RN 6148-64-7

IUPAC Name: 3-ethoxy-3-oxopropanoic acid potassium
Synonyms: ethyl hydrogen malonate potassium
Molecular Weight (g/mol): 171.21
Molecular Formula: C5H8KO4
InChi Key: HRBCMCJOBRIIQG-UHFFFAOYSA-N
SMILES: [K].CCOC(=O)CC(O)=O
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results