CAS RN 615-50-9
IUPAC Name:
2-methylbenzene-1,4-diamine; sulfuric acid
Synonyms:
toluene-2,5-diamine sulfate
Molecular Weight (g/mol):
220.24
Molecular Formula:
C7H12N2O4S
InChi Key:
KZTWOUOZKZQDMN-UHFFFAOYSA-N
SMILES:
OS(O)(=O)=O.CC1=CC(N)=CC=C1N
1
–
2
of
2
results
1
2,5-Diaminotoluene sulfate, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
2,5-Diaminotoluene Sulfate, TRC
CAS: 615-50-9 Molecular Formula: C7 H10 N2 . H2 O4 S Molecular Weight (g/mol): 220.25 Synonym: Toluene-2,5-diamine, sulfate (1:1) (8CI),2,5-Diaminotoluene sulfate,C.I. 76043,NSC 1703 IUPAC Name: 2-methylbenzene-1,4-diamine;sulfuric acid SMILES: Cc1cc(N)ccc1N.OS(=O)(=O)O