missing translation for 'onlineSavingsMsg'
Learn More

2,5-Diaminotoluene Sulfate, TRC

Product Code. 30137326
Change view
Click to view available options
Quantity:
25 g
50 g
250 g
Unit Size:
250g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137326 50 g 50g
30135718 250 g 250g
30009753 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137326 Supplier Toronto Research Chemicals Supplier No. TRCD41670550G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-50-9
Molecular Formula C7 H10 N2 . H2 O4 S
Molecular Weight (g/mol) 220.25
Synonym Toluene-2,5-diamine, sulfate (1:1) (8CI), 2,5-Diaminotoluene sulfate, C.I. 76043, NSC 1703
IUPAC Name 2-methylbenzene-1,4-diamine;sulfuric acid
SMILES Cc1cc(N)ccc1N.OS(=O)(=O)O

Specifications

Quantity 50 g
InChI Formula InChI=1S/C7H10N2.H2O4S/c1-5-4-6(8)2-3-7(5)9;1-5(2,3)4/h2-4H,8-9H2,1H3;(H2,1,2,3,4)
Formula Weight 220.0518
Chemical Name or Material 2,5-Diaminotoluene sulfate (2-Methyl-p-phenylenediamine sulfate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.