accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOBOHS

CAS RN 6165-68-0

IUPAC Name: (thiophen-2-yl)boronic acid
Synonyms: thiophen-2-ylboronic acid
Molecular Weight (g/mol): 127.95
Molecular Formula: C4H5BO2S
InChi Key: ARYHTUPFQTUBBG-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CS1
Percent Purity 
  • (4)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results