missing translation for 'onlineSavingsMsg'
Learn More

2-Thienylboronic acid, 97%, Maybridge

Product Code. 10376464
Change view
Click to view available options
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10376464 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10376464 Supplier Thermo Scientific Maybridge Supplier No. CC13012EA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 6165-68-0
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C4H5BO2S
MDL Number MFCD00151850
Quantity 10 g
Synonym thiophene-2-boronic acid, 2-thiopheneboronic acid, 2-thienylboronic acid, thienylboronic acid, 2-thiopheneboronicacid, thiophen-2-ylboranediol, thiophene2-boronic acid, thiophen-2-yl boronic acid, 2-thiophenylboric acid, boronic acid, thienyl
InChI Key ARYHTUPFQTUBBG-UHFFFAOYSA-N
SMILES OB(O)C1=CC=CS1
IUPAC Name (thiophen-2-yl)boronic acid
Molecular Weight (g/mol) 127.95
PubChem CID 2733960
Formula Weight 127.96
Percent Purity 97%
Chemical Name or Material 2-Thienylboronic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.