accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOCH3O

CAS RN 6175-45-7

IUPAC Name: 2,2-diethoxy-1-phenylethan-1-one
Synonyms: 2,2-diethoxyacetophenone
Molecular Weight (g/mol): 208.26
Molecular Formula: C12H16O3
InChi Key: PIZHFBODNLEQBL-UHFFFAOYSA-N
SMILES: CCOC(OCC)C(=O)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results