accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OONOONOO

CAS RN 618-71-3

IUPAC Name: ethyl 3,5-dinitrobenzoate
Synonyms: ethyl 3,5-dinitrobenzoate
Molecular Weight (g/mol): 240.17
Molecular Formula: C9H8N2O6
InChi Key: IBQREHJPMPCXQA-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results