CAS RN 61837-65-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(+/-)-4-Hydroxy Mephenytoin, TRC
CAS: 61837-65-8 Molecular Formula: C12H14N2O3 Molecular Weight (g/mol): 234.25 Synonym: 2,4-Imidazolidinedione, 5-ethyl-5-(4-hydroxyphenyl)-3-methyl-,5-Ethyl-5-(4-hydroxyphenyl)-3-methyl-2,4-imidazolidinedione,(±)-4-Hydroxymephenytoin,4-Hydroxymephenytoin,4'-Hydroxymephenytoin,p-Hydroxymephenytoin,(±)-4-Hydroxy mephenytoin IUPAC Name: 5-ethyl-5-(4-hydroxyphenyl)-3-methylimidazolidine-2,4-dione SMILES: CCC1(NC(=O)N(C)C1=O)c2ccc(O)cc2