missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-4-Hydroxy Mephenytoin, TRC

Product Code. 30005937
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30005937 25 mg 25mg
17951925 5 mg 5mg
17931815 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005937 Supplier Toronto Research Chemicals Supplier No. TRCH94487525MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61837-65-8
Molecular Formula C12H14N2O3
Molecular Weight (g/mol) 234.25
Synonym 2,4-Imidazolidinedione, 5-ethyl-5-(4-hydroxyphenyl)-3-methyl-, 5-Ethyl-5-(4-hydroxyphenyl)-3-methyl-2,4-imidazolidinedione, (±)-4-Hydroxymephenytoin, 4-Hydroxymephenytoin, 4'-Hydroxymephenytoin, p-Hydroxymephenytoin, (±)-4-Hydroxy mephenytoin
IUPAC Name 5-ethyl-5-(4-hydroxyphenyl)-3-methylimidazolidine-2,4-dione
SMILES CCC1(NC(=O)N(C)C1=O)c2ccc(O)cc2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C12H14N2O3/c1-3-12(8-4-6-9(15)7-5-8)10(16)14(2)11(17)13-12/h4-7,15H,3H2,1-2H3,(H,13,17)
Formula Weight 234.1004
Percent Purity >95%
Chemical Name or Material 4-Hydroxymephenytoin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.