CAS RN 619-41-0
2-Bromo-4'-methylacetophenone, 97%
CAS: 619-41-0 Molecular Formula: C9H9BrO Molecular Weight (g/mol): 213.07 MDL Number: MFCD00000203 InChI Key: KRVGXFREOJHJAX-UHFFFAOYSA-N Synonym: 2-bromo-4'-methylacetophenone,2-bromo-1-p-tolyl ethanone,4-methylphenacyl bromide,2-bromo-1-4-methylphenyl ethanone,2-bromo-1-p-tolylethanone,2-bromo-4-methylacetophenone,2-bromo-1-4-methylphenyl ethan-1-one,acetophenone, 2-bromo-4'-methyl,ethanone, 2-bromo-1-4-methylphenyl,p-methylphenacyl bromide PubChem CID: 69272 IUPAC Name: 2-bromo-1-(4-methylphenyl)ethanone SMILES: CC1=CC=C(C=C1)C(=O)CBr
2-Bromo-4'-methylacetophenone, 98%
CAS: 619-41-0 Molecular Formula: C9H9BrO Molecular Weight (g/mol): 213.074 MDL Number: MFCD00000203 InChI Key: KRVGXFREOJHJAX-UHFFFAOYSA-N Synonym: 2-bromo-4'-methylacetophenone,2-bromo-1-p-tolyl ethanone,4-methylphenacyl bromide,2-bromo-1-4-methylphenyl ethanone,2-bromo-1-p-tolylethanone,2-bromo-4-methylacetophenone,2-bromo-1-4-methylphenyl ethan-1-one,acetophenone, 2-bromo-4'-methyl,ethanone, 2-bromo-1-4-methylphenyl,p-methylphenacyl bromide PubChem CID: 69272 IUPAC Name: 2-bromo-1-(4-methylphenyl)ethanone SMILES: CC1=CC=C(C=C1)C(=O)CBr
2-Bromo-1-(4-methylphenyl)ethanone, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-Bromo-4’-methylacetophenone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2-Bromo-1-(4-methylphenyl)ethan-1-one, 97%, Maybridge™
CAS: 619-41-0 Molecular Formula: C9H9BrO Molecular Weight (g/mol): 213.074 InChI Key: KRVGXFREOJHJAX-UHFFFAOYSA-N Synonym: 2-bromo-4'-methylacetophenone,2-bromo-1-p-tolyl ethanone,4-methylphenacyl bromide,2-bromo-1-4-methylphenyl ethanone,2-bromo-1-p-tolylethanone,2-bromo-4-methylacetophenone,2-bromo-1-4-methylphenyl ethan-1-one,acetophenone, 2-bromo-4'-methyl,ethanone, 2-bromo-1-4-methylphenyl,p-methylphenacyl bromide PubChem CID: 69272 IUPAC Name: 2-bromo-1-(4-methylphenyl)ethanone SMILES: CC1=CC=C(C=C1)C(=O)CBr