CAS RN 620-92-8
IUPAC Name:
4-[(4-hydroxyphenyl)methyl]phenol
Synonyms:
bis(4-hydroxyphenyl)methane HDM 4,4'-dihydroxydiphenylmethane 4,4'-methylene diphenol p,p'-bis(hydroxyphenyl)methane
Molecular Weight (g/mol):
200.24
Molecular Formula:
C13H12O2
InChi Key:
PXKLMJQFEQBVLD-UHFFFAOYSA-N
SMILES:
OC1=CC=C(CC2=CC=C(O)C=C2)C=C1