accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 621-59-0

H3COOHO

CAS RN 621-59-0

IUPAC Name: 3-hydroxy-4-methoxybenzaldehyde
Synonyms: isovanillin
Molecular Weight (g/mol): 152.15
Molecular Formula: C8H8O3
InChi Key: JVTZFYYHCGSXJV-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=O)C=C1O
Molecular Weight (g/mol) 
  • (4)
  • (5)
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (1)
  • (3)
  • (3)
Percent Purity 
  • (1)
  • (6)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (4)
  • (5)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (6)
  • (1)
  • (3)