accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 622-78-6

IUPAC Name: (isothiocyanatomethyl)benzene
Synonyms: benzyl isothiocyanate tromacaps tromalyt urogran benzylsenfoel
Molecular Weight (g/mol): 149.21
Molecular Formula: C8H7NS
InChi Key: MDKCFLQDBWCQCV-UHFFFAOYSA-N
SMILES: S=C=NCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (3)
  • (1)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)