Learn More
Benzyl Isothiocyanate, TRC
Product Code. 30134928
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134928 50 g 50g
30005009 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30134928 Supplier Toronto Research Chemicals Supplier No. TRCB28150050G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 622-78-6
Molecular Formula C8 H7 N S
Molecular Weight (g/mol) 149.21
Synonym Isothiocyanic Acid Benzyl Ester, (Isothiocyanatomethyl)benzene, 1-(Isothiocyanatomethyl)benzene, AB 2, Benzyl Mustard Oil, NSC 118976, Phenylmethyl Isothiocyanate, Tromacaps, Tromalyt, Urogran;
IUPAC Name isothiocyanatomethylbenzene
SMILES S=C=NCc1ccccc1
Specifications
Quantity 50 g
InChI Formula InChI=1S/C8H7NS/c10-7-9-6-8-4-2-1-3-5-8/h1-5H,6H2
Formula Weight 149.0299
Chemical Name or Material Benzyl Isothiocyanate