accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COCl

CAS RN 623-12-1

IUPAC Name: 1-chloro-4-methoxybenzene
Synonyms: 4-chloroanisole
Molecular Weight (g/mol): 142.58
Molecular Formula: C7H7ClO
InChi Key: YRGAYAGBVIXNAQ-UHFFFAOYSA-N
SMILES: COC1=CC=C(Cl)C=C1
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)