missing translation for 'onlineSavingsMsg'
Learn More

4-Chloroanisole, 99%

Product Code. 15770819 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15770819 100 g Glass bottle 100g
10462780 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15770819 Supplier Thermo Scientific Acros Supplier No. 170151000
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 623-12-1 | C7H7ClO | 142.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-12-1
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.58
InChI Key YRGAYAGBVIXNAQ-UHFFFAOYSA-N
Synonym 4-chloroanisole, benzene, 1-chloro-4-methoxy, p-chloroanisole, p-chlorophenyl methyl ether, anisyl chloride, anisole, p-chloro, p-chloromethoxybenzene, 4-chlorophenol methyl ether, 1-chloro-4-methoxy-benzene, unii-0f18bly08q
PubChem CID 12167
IUPAC Name 1-chloro-4-methoxybenzene
SMILES COC1=CC=C(C=C1)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -18.0°C
Color Colorless to Yellow
Density 1.1600g/mL
Boiling Point 198.0°C to 202.0°C
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H4OCH3
Refractive Index 1.5340 to 1.5360
Quantity 100 g
Beilstein 06,186
Specific Gravity 1.16
Solubility Information Solubility in water: immiscible.
Formula Weight 142.58
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Chloroanisole, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.