accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 624-95-3

CH3CH3CH3HO

CAS RN 624-95-3

IUPAC Name: 3,3-dimethylbutan-1-ol
Synonyms: 3,3-dimethyl-1-butanol
Molecular Weight (g/mol): 102.18
Molecular Formula: C6H14O
InChi Key: DUXCSEISVMREAX-UHFFFAOYSA-N
SMILES: CC(C)(C)CCO
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results