accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3O(Z)

CAS RN 625-33-2

IUPAC Name: (3Z)-pent-3-en-2-one
Synonyms: (3Z)-pent-3-en-2-one
Molecular Weight (g/mol): 84.12
Molecular Formula: C5H8O
InChi Key: LABTWGUMFABVFG-ARJAWSKDSA-N
SMILES: C\C=C/C(C)=O
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (3)
  • (1)
  • (3)
Boiling Point 
  • (2)
  • (3)
Grade 
  • (5)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (5)
  • (3)
  • (1)
  • (3)
  • (2)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)