CAS RN 62666-20-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Progabide, TRC
CAS: 62666-20-0 Molecular Formula: C17 H16 Cl F N2 O2 Molecular Weight (g/mol): 334.77 Synonym: Butanamide, 4-[[(4-chlorophenyl)(5-fluoro-2-hydroxyphenyl)methylene]amino]-,4-[[(4-Chlorophenyl)(5-fluoro-2-hydroxyphenyl)methylene]amino]butanamide,Gabren,Gabrene,Halogabide,Progabide,SL 76-002,SL 76002 IUPAC Name: 4-[(Z)-[(4-chlorophenyl)-(5-fluoro-2-hydroxyphenyl)methylidene]amino]butanamide SMILES: NC(=O)CCC\N=C(\c1ccc(Cl)cc1)/c2cc(F)ccc2O