accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOHO

CAS RN 627-03-2

IUPAC Name: 2-ethoxyacetic acid
Synonyms: acetic acid, ethoxy-
Molecular Weight (g/mol): 104.11
Molecular Formula: C4H8O3
InChi Key: YZGQDNOIGFBYKF-UHFFFAOYSA-N
SMILES: CCOCC(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)