accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 628-12-6

H3COOClO

CAS RN 628-12-6

IUPAC Name: 2-methoxyethyl carbonochloridate
Synonyms: 2-methoxyethyl carbonochloridate
Molecular Weight (g/mol): 138.55
Molecular Formula: C4H7ClO3
InChi Key: NDYYWMXJZWHRLZ-UHFFFAOYSA-N
SMILES: COCCOC(Cl)=O
Boiling Point 
  • (3)
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)