accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OHH3CHC

CAS RN 6285-06-9

IUPAC Name: 3-ethylpent-1-yn-3-ol
Synonyms: 1-pentyn-3-ol, 3-ethyl-
Molecular Weight (g/mol): 112.17
Molecular Formula: C7H12O
InChi Key: PUNRPAWKFTXZIW-UHFFFAOYSA-N
SMILES: CCC(O)(CC)C#C
Boiling Point 
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results