accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 6289-46-9

IUPAC Name: 1,4-dimethyl 2,5-dioxocyclohexane-1,4-dicarboxylate
Synonyms: 1,4-dimethyl 2,5-dioxocyclohexane-1,4-dicarboxylate
Molecular Weight (g/mol): 228.2
Molecular Formula: C10H12O6
InChi Key: MHKKFFHWMKEBDW-UHFFFAOYNA-N
SMILES: COC(=O)C1CC(=O)C(CC1=O)C(=O)OC
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results