accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3

CAS RN 629-50-5

IUPAC Name: tridecane
Synonyms: tridecane tridekan N-tridecane
Molecular Weight (g/mol): 184.37
Molecular Formula: C13H28
InChi Key: IIYFAKIEWZDVMP-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCC
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results