missing translation for 'onlineSavingsMsg'
Learn More

Tridecane, TRC

Product Code. 30426437
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30426437 1 g 1g
30424731 5 g -
30422232 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30426437 Supplier Toronto Research Chemicals Supplier No. TRCT0781451G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-50-5
Molecular Formula C13 H28
Molecular Weight (g/mol) 184.36
Synonym Tridecane, Cactus Normal Paraffin N 13, N 13, N 13 (paraffin), NSC 66205, Paraffin N 13, n-Tridecane
IUPAC Name tridecane
SMILES CCCCCCCCCCCCC

Specifications

Quantity 1 g
InChI Formula InChI=1S/C13H28/c1-3-5-7-9-11-13-12-10-8-6-4-2/h3-13H2,1-2H3
Formula Weight 184.2191
Chemical Name or Material n-Tridecane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.