CAS RN 629-59-4
IUPAC Name:
tetradecane
Synonyms:
tetradecane tetradekan ccris 715 N-tetradecane
Molecular Weight (g/mol):
198.39
Molecular Formula:
C14H30
InChi Key:
BGHCVCJVXZWKCC-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCC
1
–
3
of
3
results
1
Tetradecane, 99%
CAS: 629-59-4 InChI Key: BGHCVCJVXZWKCC-UHFFFAOYSA-N Synonym: n-tetradecane,alkanes, c14-16,tridecane, methyl,ccris 715,tetradecane, analytical standard,alkanes, c14-30,myristyl,tetradekan,n-teradecane,olefine PubChem CID: 12389 ChEBI: CHEBI:41253 IUPAC Name: tetradecane SMILES: CCCCCCCCCCCCCC
2
(±)-Zopiclone-d3 (N-methyl-d3), CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.