accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3

CAS RN 629-94-7

IUPAC Name: henicosane
Synonyms: heneicosane
Molecular Weight (g/mol): 296.58
Molecular Formula: C21H44
InChi Key: FNAZRRHPUDJQCJ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCC
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (7)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)