missing translation for 'onlineSavingsMsg'
Learn More

TRPH Standard 1017 500 μg/mL in Hexane, Dr. Ehrenstorfer

Product Code. 30321120
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30321120 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30321120 Supplier Dr. Ehrenstorfer Supplier No. DREGA09001017HE
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Heneicosane∣n-Decane∣n-Docosane∣n-Dodecane∣n-Dotriacontane∣n-Eicosane∣n-Hentriacontane∣n-Heptacosane∣n-Heptadecane∣n-Heptatriacontane∣n-Hexacosane∣n-Hexadecane∣n-Hexatriacontane∣n-Nonacosane∣n-Nonadecane∣n-Nonatriacontane∣n-Octadecane∣n-Octatriacontane∣n-Pentacosane∣n-Pentadecane∣n-Pentatriacontane∣n-Tetradecane∣n-Tetratriacontane∣n-Tricosane∣n-Tridecane∣n-Tritriacontane∣n-Undecane∣Octacosane∣Tetracosane∣Triacontane
CAS 629-94-7, 124-18-5, 629-97-0, 112-40-3, 544-85-4, 112-95-8
Analyte or Component Names Heneicosane∣n-Decane∣n-Docosane∣n-Dodecane∣n-Dotriacontane∣n-Eicosane∣n-Hentriacontane∣n-Heptacosane∣n-Heptadecane∣n-Heptatriacontane∣n-Hexacosane∣n-Hexadecane∣n-Hexatriacontane∣n-Nonacosane∣n-Nonadecane∣n-Nonatriacontane∣n-Octadecane∣n-Octatriacontane∣n-Pentacosane∣n-Pentadecane∣n-Pentatriacontane∣n-Tetradecane∣n-Tetratriacontane∣n-Tricosane∣n-Tridecane∣n-Tritriacontane∣n-Undecane∣Octacosane∣Tetracosane∣Triacontane
Shipping Condition Ice pack (-20°C)
Certifications/Compliance ISO 17034
Solvent or Matrix Hexane
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Heneicosane 500 μg/mL∣n-Decane 500 μg/mL∣n-Docosane 500 μg/mL∣n-Dodecane 500 μg/mL∣n-Dotriacontane 500 μg/mL∣n-Eicosane 500 μg/mL∣n-Hentriacontane 500 μg/mL∣n-Heptacosane 500 μg/mL∣n-Heptadecane 500 μg/mL∣n-Heptatriacontane 500 μg/mL∣n-Hexacosane 500 μg/mL∣n-Hexadecane 500 μg/mL∣n-Hexatriacontane 500 μg/mL∣n-Nonacosane 500 μg/mL∣n-Nonadecane 500 μg/mL∣n-Nonatriacontane 500 μg/mL∣n-Octadecane 500 μg/mL∣n-Octatriacontane 500 μg/mL∣n-Pentacosane 500 μg/mL∣n-Pentadecane 500 μg/mL∣n-Pentatriacontane 500 μg/mL∣n-Tetradecane 500 μg/mL∣n-Tetratriacontane 500 μg/mL∣n-Tricosane 500 μg/mL∣n-Tridecane 500 μg/mL∣n-Tritriacontane 500 μg/mL∣n-Undecane 500 μg/mL∣Octacosane 500 μg/mL∣Tetracosane 500 μg/mL∣Triacontane 500 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.