accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 630-03-5

IUPAC Name: nonacosane
Synonyms: nonacosane
Molecular Weight (g/mol): 408.8
Molecular Formula: C29H60
InChi Key: IGGUPRCHHJZPBS-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCCCCCCCC
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results