missing translation for 'onlineSavingsMsg'
Learn More

Nonacosane -, TRC

Product Code. 30473350
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30473350 100 mg 100mg
30466442 250 mg 250mg
30465320 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30473350 Supplier Toronto Research Chemicals Supplier No. TRCN231970100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 630-03-5
Molecular Formula C29 H60
Molecular Weight (g/mol) 408.79
Synonym Nonacosane, Celidoniol, deoxy- (7CI), n-Nonacosane
IUPAC Name nonacosane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C29H60/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-29H2,1-2H3
Formula Weight 408.4695
Chemical Name or Material n-Nonacosane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.