accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 630-19-3

IUPAC Name: 2,2-dimethylpropanal
Synonyms: pivaldehyde pivalaldehyde
Molecular Weight (g/mol): 86.13
Molecular Formula: C5H10O
InChi Key: FJJYHTVHBVXEEQ-UHFFFAOYSA-N
SMILES: CC(C)(C)C=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
Boiling Point 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results