accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 630-20-6

ClClClCl

CAS RN 630-20-6

IUPAC Name: 1,1,1,2-tetrachloroethane
Synonyms: tetrachloroethane 1,1,1,2-tetrachloraethan C2H2cl4 ccris 577 rcra waste number U208
Molecular Weight (g/mol): 167.84
Molecular Formula: C2H2Cl4
InChi Key: QVLAWKAXOMEXPM-UHFFFAOYSA-N
SMILES: ClCC(Cl)(Cl)Cl
Molecular Weight (g/mol) 
  • (3)
  • (3)
Solvent or Matrix 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Concentration 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)