missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 8010 Halogenated VOC Mixture 2 2000 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30309644
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30309644 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30309644 Supplier Dr. Ehrenstorfer Supplier No. DREYA08010200ME
Big Savings for Better Separation
Use promo code 27992
View all promotions
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,1,1,2-Tetrachloroethane∣1,1,1-Trichloroethane∣1,1,2,2-Tetrachloroethane∣1,1,2-Trichloroethane∣1,1-Dichloroethane∣1,2,3-Trichloropropane∣1,2-Dichloroethane∣1,2-Dichloropropane∣Chloroform∣Dibromomethane∣Tetrachloroethene∣Tetrachloromethane∣Tribromomethane∣Trichloroethene
CAS 630-20-6, 71-55-6, 79-34-5, 79-00-5, 75-34-3, 96-18-4
Analyte or Component Names 1,1,1,2-Tetrachloroethane∣1,1,1-Trichloroethane∣1,1,2,2-Tetrachloroethane∣1,1,2-Trichloroethane∣1,1-Dichloroethane∣1,2,3-Trichloropropane∣1,2-Dichloroethane∣1,2-Dichloropropane∣Chloroform∣Dibromomethane∣Tetrachloroethene∣Tetrachloromethane∣Tribromomethane∣Trichloroethene
Shipping Condition Room Temperature
Certifications/Compliance ISO/IEC 17025
Solution Type Mixture
Solvent or Matrix Methanol
Industry Type Environmental Testing
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 1,1,1,2-Tetrachloroethane 2000 μg/mL∣1,1,1-Trichloroethane 2000 μg/mL∣1,1,2,2-Tetrachloroethane 2000 μg/mL∣1,1,2-Trichloroethane 2000 μg/mL∣1,1-Dichloroethane 2000 μg/mL∣1,2,3-Trichloropropane 2000 μg/mL∣1,2-Dichloroethane 2000 μg/mL∣1,2-Dichloropropane 2000 μg/mL∣Chloroform 2000 μg/mL∣Dibromomethane 2000 μg/mL∣Tetrachloroethene 2000 μg/mL∣Tetrachloromethane 2000 μg/mL∣Tribromomethane 2000 μg/mL∣Trichloroethene 2000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.