accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 63131-29-3

IUPAC Name: methyl 3-(4-fluorophenyl)-3-oxopropanoate
Synonyms: methyl 3-(4-fluorophenyl)-3-oxopropanoate
Molecular Weight (g/mol): 196.18
Molecular Formula: C10H9FO3
InChi Key: HGLVYXXPRSNKQN-UHFFFAOYSA-N
SMILES: COC(=O)CC(=O)C1=CC=C(F)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)