accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 6342-56-9

H3COOCH3CH3O

CAS RN 6342-56-9

IUPAC Name: 1,1-dimethoxypropan-2-one
Synonyms: 1,1-dimethoxypropan-2-one
Molecular Weight (g/mol): 118.13
Molecular Formula: C5H10O3
InChi Key: ULVSHNOGEVXRDR-UHFFFAOYSA-N
SMILES: COC(OC)C(C)=O
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)